siRNA Details
| Gene Symbol: Echs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078623.2 | NP_511178.1 | 6 | 654 | ||
| GenBank mRNA | BC064655.1 | - | 6 | 654 | 5599860 | |
| FQ218439.1 | - | 5 | 513 | |||
| FQ218627.1 | - | 3 | 198 | |||
| X15958.1 | - | 6 | 648 | |||
| Gene Symbol: LOC100911186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748886.2 | XP_003748934.1 | 6 | 682 | ||

Echs1
Genomic Map