siRNA Details
| Gene Symbol: Chst10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080397.1 | NP_536322.1 | 5 | 720 | ||
| XM_006244730.2 | XP_006244792.1 | 6 | 869 | |||
| XM_006244732.2 | XP_006244794.1 | 5 | 527 | |||
| XM_008766985.1 | XP_008765207.1 | 5 | 651 | |||
| XM_008766987.1 | XP_008765209.1 | 5 | 621 | |||
| GenBank mRNA | AF022729.1 | - | 5 | 720 | ||

Chst10
Genomic Map