siRNA Details
| Gene Symbol: Gli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191910.1 | NP_001178839.1 | 6 | 740 | ||
| XM_006241442.2 | XP_006241504.1 | 8 | 1005 | |||
| XM_006241443.2 | XP_006241505.1 | 7 | 888 | |||
| XM_008765375.1 | XP_008763597.1 | 8 | 953 | |||

Gli1
Genomic Map