siRNA Details
| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080407.1 | NP_536332.1 | 12 | 2126 | ||
| XM_006244951.2 | XP_006245013.1 | 13 | 2612 | |||
| XM_006244952.2 | XP_006245014.1 | 13 | 2612 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmpr2| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080407.1 | NP_536332.1 | 12 | 2126 | ||
| XM_006244951.2 | XP_006245013.1 | 13 | 2612 | |||
| XM_006244952.2 | XP_006245014.1 | 13 | 2612 | |||