siRNA Details
| Gene Symbol: Ppfia4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080409.1 | NP_536334.1 | 24 | 3116 | ||
| XM_008769527.1 | XP_008767749.1 | 24 | 3378 | |||
| XM_008769528.1 | XP_008767750.1 | 23 | 3351 | |||
| XM_008769529.1 | XP_008767751.1 | 24 | 3378 | |||
| XM_008769530.1 | XP_008767752.1 | 23 | 3976 | |||
| GenBank mRNA | AY057064.1 | - | 21 | 2407 | ||
| BC092640.1 | - | 5 | 381 | |||

Ppfia4
Genomic Map