siRNA Details
| Gene Symbol: Siah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134457.1 | NP_604452.1 | 2 | 932 | ||
| GenBank mRNA | AB067815.1 | - | 2 | 932 | ||
| AF389477.1 | - | 2 | 486 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Siah2| Gene Symbol: Siah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134457.1 | NP_604452.1 | 2 | 932 | ||
| GenBank mRNA | AB067815.1 | - | 2 | 932 | ||
| AF389477.1 | - | 2 | 486 | |||