siRNA Details
Gene Symbol: Rps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009239.1 | NP_001009239.1 | 6 | 643 | ||
GenBank mRNA | BC088450.1 | - | 6 | 643 | 7126819 | |
X51536.1 | - | 6 | 649 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009239.1 | NP_001009239.1 | 6 | 643 | ||
GenBank mRNA | BC088450.1 | - | 6 | 643 | 7126819 | |
X51536.1 | - | 6 | 649 |