siRNA Details
| Gene Symbol: Rps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009239.1 | NP_001009239.1 | 2 | 149 | ||
| GenBank mRNA | BC088450.1 | - | 2 | 149 | 7126819 | |
| X51536.1 | - | 2 | 155 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps3| Gene Symbol: Rps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009239.1 | NP_001009239.1 | 2 | 149 | ||
| GenBank mRNA | BC088450.1 | - | 2 | 149 | 7126819 | |
| X51536.1 | - | 2 | 155 | |||