siRNA Details
| Gene Symbol: Rps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169146.1 | NP_001162617.1 | 5 | 615 | ||
| GenBank mRNA | BC159438.1 | - | 5 | 586 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps16| Gene Symbol: Rps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169146.1 | NP_001162617.1 | 5 | 615 | ||
| GenBank mRNA | BC159438.1 | - | 5 | 586 | ||