siRNA Details
| Gene Symbol: A1bg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022258.2 | NP_071594.2 | 3 | 262 | ||
| GenBank mRNA | AJ302031.1 | - | 3 | 262 | ||
| BC089771.1 | - | 3 | 245 | 7326280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A1bg| Gene Symbol: A1bg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022258.2 | NP_071594.2 | 3 | 262 | ||
| GenBank mRNA | AJ302031.1 | - | 3 | 262 | ||
| BC089771.1 | - | 3 | 245 | 7326280 | ||