siRNA Details
| Gene Symbol: Rab3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080580.2 | NP_542147.1 | 3, 4 | 467 | ||
| XM_006242593.1 | XP_006242655.1 | 3, 4 | 463 | |||
| XM_006242594.2 | XP_006242656.1 | 3, 4 | 583 | |||
| GenBank mRNA | BC081741.1 | - | 3, 4 | 467 | 7110109 | |
| M83681.1 | - | 2, 3 | 295 | |||
| U90206.1 | - | 2, 3 | 358 | |||

Rab3d
Genomic Map