siRNA Details
| Gene Symbol: Ap3m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133305.2 | NP_579839.1 | 8 | 1318 | ||
| GenBank mRNA | BC086993.1 | - | 8 | 1318 | 7131933 | |
| L07074.1 | - | 1141 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap3m2| Gene Symbol: Ap3m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133305.2 | NP_579839.1 | 8 | 1318 | ||
| GenBank mRNA | BC086993.1 | - | 8 | 1318 | 7131933 | |
| L07074.1 | - | 1141 | ||||