siRNA Details
| Gene Symbol: Abcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080581.1 | NP_542148.1 | 10 | 1243 | ||
| GenBank mRNA | AB010467.1 | - | 10 | 1243 | ||
| AF072816.1 | - | 10 | 1255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcc3| Gene Symbol: Abcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080581.1 | NP_542148.1 | 10 | 1243 | ||
| GenBank mRNA | AB010467.1 | - | 10 | 1243 | ||
| AF072816.1 | - | 10 | 1255 | |||