siRNA Details
| Gene Symbol: Abcb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080582.1 | NP_542149.1 | 14 | 2099 | ||
| GenBank mRNA | AF106563.1 | - | 6 | 486 | ||
| AJ003004.1 | - | 14 | 2099 | |||
| BC085712.1 | - | 14 | 2115 | 7190755 | ||
| FQ211570.1 | - | 14 | 2125 | |||

Abcb6
Genomic Map