siRNA Details
| Gene Symbol: Ap2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080583.1 | NP_542150.1 | 6 | 837 | ||
| XM_008767935.1 | XP_008766157.1 | 7 | 1594 | |||
| XM_008767936.1 | XP_008766158.1 | 6 | 808 | |||
| XM_008767937.1 | XP_008766159.1 | 5 | 723 | |||
| XM_008767938.1 | XP_008766160.1 | 5 | 740 | |||
| XM_008767939.1 | XP_008766161.1 | 6 | 1576 | |||
| GenBank mRNA | BC103481.1 | - | 7 | 806 | 7929959 | |
| M34176.1 | - | 6 | 769 | |||
| M77246.1 | - | 6 | 837 | |||
| Gene Symbol: LOC103690090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773863.1 | XP_008772085.1 | 6 | 1576 | ||
| XM_008773864.1 | XP_008772086.1 | 7 | 1594 | |||
| XM_008773865.1 | XP_008772087.1 | 6 | 808 | |||
| XM_008773866.1 | XP_008772088.1 | 5 | 723 | |||
| XM_008773867.1 | XP_008772089.1 | 6 | 740 | |||

Ap2b1
Genomic Map