siRNA Details
| Gene Symbol: Phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080584.4 | NP_542151.1 | 6 | 922 | ||
| XM_008759855.1 | XP_008758077.1 | 7 | 745 | |||
| XM_008759856.1 | XP_008758078.1 | 7 | 863 | |||
| GenBank mRNA | BC107906.1 | - | 6 | 782 | ||
| BC128715.1 | - | 6 | 851 | 7456955 | ||
| M73808.1 | - | 6 | 922 | |||

Phkg2
Genomic Map