siRNA Details
| Gene Symbol: Gabra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080587.3 | NP_542154.3 | 1, 2 | 178 | ||
| GenBank mRNA | S55933.1 | - | 1, 2 | 178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabra4| Gene Symbol: Gabra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080587.3 | NP_542154.3 | 1, 2 | 178 | ||
| GenBank mRNA | S55933.1 | - | 1, 2 | 178 | ||