siRNA Details
| Gene Symbol: Caskin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080690.2 | NP_542421.2 | 9 | 1048 | ||
| XM_006245895.2 | XP_006245957.1 | 9 | 2324 | |||
| XM_006245896.2 | XP_006245958.1 | 9 | 2324 | |||
| XM_006245897.2 | XP_006245959.1 | 9 | 2303 | |||
| GenBank mRNA | AF451975.1 | - | 9 | 1071 | ||

Caskin1
Genomic Map