siRNA Details
| Gene Symbol: Cacng7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080695.1 | NP_542426.1 | 4 | 509 | ||
| XM_006228013.2 | XP_006228075.1 | 5 | 648 | |||
| GenBank mRNA | AF361345.1 | - | 4 | 509 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacng7| Gene Symbol: Cacng7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080695.1 | NP_542426.1 | 4 | 509 | ||
| XM_006228013.2 | XP_006228075.1 | 5 | 648 | |||
| GenBank mRNA | AF361345.1 | - | 4 | 509 | ||