siRNA Details
| Gene Symbol: Cacng8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080696.2 | NP_542427.1 | 4 | 578 | ||
| GenBank mRNA | AF361346.1 | - | 4 | 578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacng8| Gene Symbol: Cacng8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080696.2 | NP_542427.1 | 4 | 578 | ||
| GenBank mRNA | AF361346.1 | - | 4 | 578 | ||