siRNA Details
| Gene Symbol: Manea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080785.2 | NP_542963.2 | 1 | 315 | ||
| XM_006237960.1 | XP_006238022.1 | 2 | 619 | |||
| XM_006237961.2 | XP_006238023.1 | 2 | 538 | |||
| GenBank mRNA | AY599499.1 | - | 1 | 315 | ||
| FQ223109.1 | - | 2 | 544 | |||

Manea
Genomic Map