siRNA Details
| Gene Symbol: Slco2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080786.1 | NP_542964.1 | 3, 4 | 342 | ||
| XM_006229721.2 | XP_006229783.1 | 3, 4 | 648 | |||
| XM_006229722.2 | XP_006229784.1 | 3, 4 | 648 | |||
| XM_006229723.2 | XP_006229785.1 | 3, 4 | 1648 | |||
| XM_006229724.2 | XP_006229786.1 | 3, 4 | 1619 | |||
| XM_006229725.2 | XP_006229787.1 | 3, 4 | 1604 | |||
| XM_006229727.2 | XP_006229789.1 | 3, 4 | 1619 | |||
| XM_006229728.2 | XP_006229790.1 | 3, 4 | 1604 | |||
| XM_008759668.1 | XP_008757890.1 | 4, 5 | 438 | |||
| GenBank mRNA | AF169409.1 | - | 3, 4 | 342 | ||
| AF169410.1 | - | 3, 4 | 302 | |||

Slco2b1
Genomic Map