siRNA Details
| Gene Symbol: Cdk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080885.1 | NP_543161.1 | 2 | 202 | ||
| GenBank mRNA | L02121.1 | - | 2 | 202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk5| Gene Symbol: Cdk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080885.1 | NP_543161.1 | 2 | 202 | ||
| GenBank mRNA | L02121.1 | - | 2 | 202 | ||