siRNA Details
Gene Symbol: Cdk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080885.1 | NP_543161.1 | 11 | 845 | ||
GenBank mRNA | L02121.1 | - | 11 | 845 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080885.1 | NP_543161.1 | 11 | 845 | ||
GenBank mRNA | L02121.1 | - | 11 | 845 |