siRNA Details
Gene Symbol: Slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177481.1 | NP_803434.1 | 2 | 496 | ||
XM_006229378.2 | XP_006229440.1 | 2 | 661 | |||
GenBank mRNA | AF239219.1 | - | 2 | 496 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177481.1 | NP_803434.1 | 2 | 496 | ||
XM_006229378.2 | XP_006229440.1 | 2 | 661 | |||
GenBank mRNA | AF239219.1 | - | 2 | 496 |