siRNA Details
| Gene Symbol: Il2rg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080889.1 | NP_543165.1 | 2, 3 | 283 | ||
| GenBank mRNA | AF410926.1 | - | 2, 3 | 203 | ||
| BC079343.1 | - | 2, 3 | 283 | 7106866 | ||
| FQ232752.1 | - | 1, 2 | 84 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il2rg| Gene Symbol: Il2rg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080889.1 | NP_543165.1 | 2, 3 | 283 | ||
| GenBank mRNA | AF410926.1 | - | 2, 3 | 203 | ||
| BC079343.1 | - | 2, 3 | 283 | 7106866 | ||
| FQ232752.1 | - | 1, 2 | 84 | |||