siRNA Details
| Gene Symbol: Selenbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080892.1 | NP_543168.1 | 10 | 1306 | ||
| GenBank mRNA | AB036799.1 | - | 10 | 1306 | ||
| BC074008.1 | - | 10 | 1316 | 7100467 | ||
| FQ219337.1 | - | 10 | 1287 | |||
| Gene Symbol: LOC103689947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761279.1 | XP_008759501.1 | 1306 | |||

Selenbp1
Genomic Map