siRNA Details
| Gene Symbol: Pde7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080894.2 | NP_543170.1 | 13 | 1315 | ||
| XM_008758591.1 | XP_008756813.1 | 12 | 1635 | |||
| XM_008758592.1 | XP_008756814.1 | 12 | 1219 | |||
| GenBank mRNA | AB057409.1 | - | 13 | 1315 | ||
| AB057410.1 | - | 12 | 1183 | |||
| AB057411.1 | - | 14 | 1284 | |||

Pde7b
Genomic Map