siRNA Details
| Gene Symbol: Hnrnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080896.1 | NP_543172.1 | 2 | 209 | ||
| GenBank mRNA | AF367468.1 | - | 2 | 209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnrnph1| Gene Symbol: Hnrnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080896.1 | NP_543172.1 | 2 | 209 | ||
| GenBank mRNA | AF367468.1 | - | 2 | 209 | ||