siRNA Details
| Gene Symbol: Trpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080898.2 | NP_543174.1 | 5 | 1447 | ||
| GenBank mRNA | AY064411.1 | - | 1778 | |||
| EF672039.1 | - | 5 | 1447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpc5| Gene Symbol: Trpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080898.2 | NP_543174.1 | 5 | 1447 | ||
| GenBank mRNA | AY064411.1 | - | 1778 | |||
| EF672039.1 | - | 5 | 1447 | |||