siRNA Details
Gene Symbol: Trpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080898.2 | NP_543174.1 | 8 | 2125 | ||
GenBank mRNA | AY064411.1 | - | 2456 | |||
EF672039.1 | - | 8 | 2125 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080898.2 | NP_543174.1 | 8 | 2125 | ||
GenBank mRNA | AY064411.1 | - | 2456 | |||
EF672039.1 | - | 8 | 2125 |