siRNA Details
Gene Symbol: Psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134458.2 | NP_604453.2 | 7 | 584 | ||
XM_006247249.2 | XP_006247311.1 | 7 | 1436 | |||
GenBank mRNA | AF352812.1 | - | 7 | 584 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psmc3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134458.2 | NP_604453.2 | 7 | 584 | ||
XM_006247249.2 | XP_006247311.1 | 7 | 1436 | |||
GenBank mRNA | AF352812.1 | - | 7 | 584 |