siRNA Details
| Gene Symbol: Arf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080904.2 | NP_543180.1 | 2, 3 | 341 | ||
| XM_006257320.2 | XP_006257382.1 | 3, 4 | 550 | |||
| GenBank mRNA | AY325223.1 | - | 3, 4 | 308 | ||
| BC088865.1 | - | 2, 3 | 341 | 7319260 | ||
| L12382.1 | - | 3, 4 | 454 | |||

Arf3
Genomic Map