siRNA Details
| Gene Symbol: Cfh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130409.2 | NP_569093.2 | 19, 20 | 3193 | ||
| XM_008769531.1 | XP_008767753.1 | 18, 19 | 3219 | |||
| XM_008769532.1 | XP_008767754.1 | 10, 11 | 1751 | |||
| GenBank mRNA | AJ320522.1 | - | 3167 | |||
| BC083174.1 | - | 1564 | 7096787 | |||
| CF110979.1 | - | 3, 4 | 549 | |||

Cfh
Genomic Map