siRNA Details
| Gene Symbol: Cfh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130409.2 | NP_569093.2 | 20, 21 | 3370 | ||
| XM_008769531.1 | XP_008767753.1 | 19, 20 | 3396 | |||
| XM_008769532.1 | XP_008767754.1 | 11, 12 | 1928 | |||
| GenBank mRNA | AJ320522.1 | - | 3347 | |||
| BC083174.1 | - | 1741 | 7096787 | |||
| CF110979.1 | - | 4, 5 | 726 | |||

Cfh
Genomic Map