siRNA Details
| Gene Symbol: Il23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130410.2 | NP_569094.1 | 3 | 602 | ||
| GenBank mRNA | AY055379.1 | - | 3 | 330 | ||
| BC098907.1 | - | 3 | 602 | 7454618 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il23a| Gene Symbol: Il23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130410.2 | NP_569094.1 | 3 | 602 | ||
| GenBank mRNA | AY055379.1 | - | 3 | 330 | ||
| BC098907.1 | - | 3 | 602 | 7454618 | ||