siRNA Details
| Gene Symbol: Sdf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130412.2 | NP_569096.2 | 4 | 576 | ||
| XM_006239516.2 | XP_006239578.1 | 4 | 624 | |||
| GenBank mRNA | AF405545.1 | - | 3 | 486 | ||
| BC086996.1 | - | 4 | 576 | 7192272 | ||
| FQ218523.1 | - | 4 | 601 | |||
| FQ219323.1 | - | 4 | 603 | |||
| FQ232476.1 | - | 4 | 599 | |||

Sdf4
Genomic Map