siRNA Details
| Gene Symbol: Abcg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130414.2 | NP_569098.2 | 10 | 1573 | ||
| XM_008764453.1 | XP_008762675.1 | 8 | 1614 | |||
| GenBank mRNA | AF351785.2 | - | 10 | 1573 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcg8| Gene Symbol: Abcg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130414.2 | NP_569098.2 | 10 | 1573 | ||
| XM_008764453.1 | XP_008762675.1 | 8 | 1614 | |||
| GenBank mRNA | AF351785.2 | - | 10 | 1573 | ||