siRNA Details
| Gene Symbol: Anxa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130416.2 | NP_569100.2 | 10 | 1180 | ||
| XM_006251624.2 | XP_006251686.1 | 11 | 1207 | |||
| XM_006251625.2 | XP_006251687.1 | 11 | 3006 | |||
| XM_006251626.2 | XP_006251688.1 | 12 | 3087 | |||
| XM_006251627.2 | XP_006251689.1 | 10 | 1139 | |||
| XM_008770484.1 | XP_008768706.1 | 11 | 1388 | |||
| XM_008770485.1 | XP_008768707.1 | 10 | 1321 | |||
| GenBank mRNA | AF280423.1 | - | 1172 | |||
| BC070896.1 | - | 10 | 1180 | 7099644 | ||

Anxa7
Genomic Map