siRNA Details
| Gene Symbol: Hey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130417.1 | NP_569101.1 | 5 | 1041 | ||
| GenBank mRNA | AY059382.1 | - | 1 | 514 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hey2| Gene Symbol: Hey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130417.1 | NP_569101.1 | 5 | 1041 | ||
| GenBank mRNA | AY059382.1 | - | 1 | 514 | ||