siRNA Details
| Gene Symbol: Trim9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130420.1 | NP_569104.1 | 2 | 1007 | ||
| XM_008764691.1 | XP_008762913.1 | 2 | 1048 | |||
| XM_008764692.1 | XP_008762914.1 | 2 | 1048 | |||
| XM_008764693.1 | XP_008762915.1 | 2 | 1048 | |||
| XM_008764694.1 | XP_008762916.1 | 2 | 1048 | |||
| GenBank mRNA | AF350422.1 | - | 2 | 1007 | ||

Trim9
Genomic Map