siRNA Details
| Gene Symbol: Casp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130422.1 | NP_569106.1 | 5 | 654 | ||
| XM_008765891.1 | XP_008764113.1 | 6 | 735 | |||
| GenBank mRNA | AF317633.1 | - | 5 | 654 | ||
| FJ465154.1 | - | 6 | 717 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casp12| Gene Symbol: Casp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130422.1 | NP_569106.1 | 5 | 654 | ||
| XM_008765891.1 | XP_008764113.1 | 6 | 735 | |||
| GenBank mRNA | AF317633.1 | - | 5 | 654 | ||
| FJ465154.1 | - | 6 | 717 | |||