siRNA Details
Gene Symbol: Casp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130422.1 | NP_569106.1 | 6, 7 | 926 | ||
XM_008765891.1 | XP_008764113.1 | 7, 8 | 1007 | |||
GenBank mRNA | AF317633.1 | - | 6, 7 | 926 | ||
FJ465154.1 | - | 7, 8 | 989 |
