siRNA Details
| Gene Symbol: Eya2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130427.1 | NP_569111.1 | 9, 10 | 960 | ||
| XM_008762456.1 | XP_008760678.1 | 10, 11 | 1196 | |||
| GenBank mRNA | AB073099.1 | - | 4, 5 | 435 | ||
| AY366465.1 | - | 9, 10 | 960 | |||

Eya2
Genomic Map