siRNA Details
| Gene Symbol: Lef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130429.1 | NP_569113.1 | 8 | 857 | ||
| GenBank mRNA | AF198533.1 | - | 8 | 857 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lef1| Gene Symbol: Lef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130429.1 | NP_569113.1 | 8 | 857 | ||
| GenBank mRNA | AF198533.1 | - | 8 | 857 | ||