siRNA Details
| Gene Symbol: Ifi27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203410.1 | NP_981955.1 | 6 | 536 | ||
| GenBank mRNA | BC060548.1 | - | 6 | 536 | 6919535 | |
| FQ219860.1 | - | 6 | 594 | |||
| FQ220098.1 | - | 6 | 605 | |||
| FQ226050.1 | - | 5 | 655 | |||
| FQ226998.1 | - | 6 | 569 | |||
| FQ230691.1 | - | 5 | 556 | |||
| FQ233730.1 | - | 6 | 570 | |||
| FQ233981.1 | - | 6 | 605 | |||

Ifi27
Genomic Map