siRNA Details
| Gene Symbol: Cygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130744.2 | NP_570100.1 | 2 | 466 | ||
| GenBank mRNA | AJ245663.2 | - | 2 | 374 | ||
| BC088455.1 | - | 2 | 466 | 7127197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cygb| Gene Symbol: Cygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130744.2 | NP_570100.1 | 2 | 466 | ||
| GenBank mRNA | AJ245663.2 | - | 2 | 374 | ||
| BC088455.1 | - | 2 | 466 | 7127197 | ||