siRNA Details
| Gene Symbol: Prkcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133307.1 | NP_579841.1 | 8 | 1065 | ||
| GenBank mRNA | AF219629.1 | - | 8 | 1059 | ||
| BC076505.1 | - | 7 | 891 | 7099952 | ||
| M18330.1 | - | 8 | 1065 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkcd| Gene Symbol: Prkcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133307.1 | NP_579841.1 | 8 | 1065 | ||
| GenBank mRNA | AF219629.1 | - | 8 | 1059 | ||
| BC076505.1 | - | 7 | 891 | 7099952 | ||
| M18330.1 | - | 8 | 1065 | |||