siRNA Details
| Gene Symbol: Prkcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133307.1 | NP_579841.1 | 11, 12 | 1446 | ||
| GenBank mRNA | AF219629.1 | - | 11, 12 | 1523 | ||
| BC076505.1 | - | 10, 11 | 1272 | 7099952 | ||
| M18330.1 | - | 11, 12 | 1446 | |||

Prkcd
Genomic Map