siRNA Details
| Gene Symbol: Slc5a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130746.1 | NP_570102.1 | 16 | 1972 | ||
| GenBank mRNA | AF026554.1 | - | 16 | 1972 | ||
| AF081204.2 | - | 13 | 1560 | |||
| AF081205.1 | - | 13 | 1560 | |||
| BC168145.1 | - | 14 | 1834 | 8371761 | ||
| FQ220013.1 | - | 2 | 69 | |||

Slc5a6
Genomic Map